Learn More
Genscript Corporation CHO-K1/OX2 Stable Cell Line
Catalog No. 5000058382
Encompass
Recombinant CHO-K1 cells stably overexpress human hypocretin (orexin) receptor 2 (OX2) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58382 Supplier Genscript Corporation Supplier No. M00316
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human hypocretin (orexin) receptor 2 (OX2) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways