Learn More
Genscript Corporation CHO-K1/OX40 Stable Cell Line
Catalog No. 5000057466
Encompass
OX40 (CD134) is a member of the TNFR/TNF superfamily Costimulatory signals from OX40 to a conventional T cell promotes division and survival augmenting the clonal expansion of effector and memory populations as they are being generated to antigens OX40 additionally suppresses the differentiation and activity of Treg further amplifying this process
Catalog No. 50-000-57466 Supplier Genscript Corporation Supplier No. M00561
Add to Cart
Add to Cart
Request a Quote
OX40 (CD134) is a member of the TNFR/TNF superfamily Costimulatory signals from OX40 to a conventional T cell promotes division and survival augmenting the clonal expansion of effector and memory populations as they are being generated to antigens OX40 additionally suppresses the differentiation and activity of Treg further amplifying this process