Learn More
Genscript Corporation CHO-K1/PAC1/G15 Stable Cell L
Catalog No. 5000058622
Encompass
Recombinant CHO-K1 cells stably overexpress human pituitary adenylate cyclase-activating polypeptide type I receptor (PAC1) on the surface and contain high levels of G protein G q and G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58622 Supplier Genscript Corporation Supplier No. M00203
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human pituitary adenylate cyclase-activating polypeptide type I receptor (PAC1) on the surface and contain high levels of G protein G q and G s to couple with the receptor in downstream signaling pathways