Learn More
Genscript Corporation CHO-K1/PD-L1 Stable Cell Line
Catalog No. 5000057306
Encompass
Recombinant CHO-K1 cells stably overexpress human programmed cell death 1 ligand 1 (PD-L1) on the surface The surface expression of PD-L1 is validated by FACS analysis This cell line is recommended for cell-based binding assay to screen antibodies against PD-L1 or to measure binding affinity between PD-L1 and anti-PD-L1 antibodies
Catalog No. 50-000-57306 Supplier Genscript Corporation Supplier No. M00543
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human programmed cell death 1 ligand 1 (PD-L1) on the surface The surface expression of PD-L1 is validated by FACS analysis This cell line is recommended for cell-based binding assay to screen antibodies against PD-L1 or to measure binding affinity between PD-L1 and anti-PD-L1 antibodies