Learn More
Genscript Corporation CHO-K1/PRLHR Stable Cell Line
Catalog No. 5000058052
Encompass
The prolactin releasing hormone receptor PRLHR also named PrRP receptor is a G-protein coupled receptor that binds the prolactin releasing hormone RT-PCR analysis showed expression of PRLHR in the human brain pituitaries normal portions of adrenal glands and various tumor tissues Northern blot analysis showed high expression of PRLHR only in tumor tissues of pheochromocytomas indicating that PRLHR expression is high in pheochromocytomas The present study has shown that PRLHR mRNA was widely expressed in the brain tissues pituitaries adrenal glands and various tumors The high expression of PRLHR receptor in pheochromocytomas suggests potential pathophysiological roles of PRLHR in these tumors
Catalog No. 50-000-58052 Supplier Genscript Corporation Supplier No. M00341
Add to Cart
Add to Cart
Request a Quote
The prolactin releasing hormone receptor PRLHR also named PrRP receptor is a G-protein coupled receptor that binds the prolactin releasing hormone RT-PCR analysis showed expression of PRLHR in the human brain pituitaries normal portions of adrenal glands and various tumor tissues Northern blot analysis showed high expression of PRLHR only in tumor tissues of pheochromocytomas indicating that PRLHR expression is high in pheochromocytomas The present study has shown that PRLHR mRNA was widely expressed in the brain tissues pituitaries adrenal glands and various tumors The high expression of PRLHR receptor in pheochromocytomas suggests potential pathophysiological roles of PRLHR in these tumors