Learn More
Genscript Corporation CHO-K1/PTAFR Stable Cell Line
Catalog No. 5000059046
Encompass
Recombinant CHO-K1 cells stably overexpress human platelet activating factor receptor (PTAFR) on the surface and contain high levels of G protein G q and G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59046 Supplier Genscript Corporation Supplier No. M00262
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human platelet activating factor receptor (PTAFR) on the surface and contain high levels of G protein G q and G i to couple with the receptor in downstream signaling pathways