Learn More
Genscript Corporation CHO-K1/Spike Stable Cell Line
Supplier:  Genscript Corporation M00803
Encompass
Recombinant CHO-K1 cells stably overexpress human SARS-CoV-2 spike protein on their surface The surface expression of SARS-CoV-2 spike protein is validated by FACS analysis This stable cell line product is designed for screening antibodies against the SARS-CoV-2 spike protein as well as measuring binding affinity and stability of antibody-based biologics that bind with spike protein GenScript also offers spike protein expressing HEK293T stable cell line (Cat No M00804) for SARS-CoV-2 study
Catalog No. 50-000-56740
Add to Cart
Request a Quote