Learn More
Genscript Corporation CHO-K1/SST2/G15 Stable Cell L
Catalog No. 5000057863
Encompass
Recombinant CHO-K1 cells stably overexpress human somatostatin receptor 2 (SST2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57863 Supplier Genscript Corporation Supplier No. M00325
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human somatostatin receptor 2 (SST2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways