Learn More
Genscript Corporation CHO-K1/SST4/G15 Stable Cell L
Catalog No. 5000057385
Encompass
Recombinant CHO-K1 cells stably overexpress human somatostatin receptor 4 (SST4) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57385 Supplier Genscript Corporation Supplier No. M00334
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human somatostatin receptor 4 (SST4) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways