Learn More
Genscript Corporation CHO-K1/TRH1 Stable Cell Line
Catalog No. 5000057386
Encompass
Thyrtropin-releasing hormone receptor1 (TRH1) is a member of G-protein coupled receptor family A This protein is a receptor for Thyrtropin-releasing hormone (TRH) Human TRH1 is expressed in lymphocytes pituitary gland and CNS It can stimulate the releasing of prolactin (PRL) thyrotropin (TSH) TRH1 receptor knockout mice exhibit a slightly reduced growth rate considerable decrease in serum T sub 3 /sub T sub 4 /sub and prolactin levels but no alteration of thyroid-stimulating hormone levels
Catalog No. 50-000-57386 Supplier Genscript Corporation Supplier No. M00202
Add to Cart
Add to Cart
Request a Quote
Thyrtropin-releasing hormone receptor1 (TRH1) is a member of G-protein coupled receptor family A This protein is a receptor for Thyrtropin-releasing hormone (TRH) Human TRH1 is expressed in lymphocytes pituitary gland and CNS It can stimulate the releasing of prolactin (PRL) thyrotropin (TSH) TRH1 receptor knockout mice exhibit a slightly reduced growth rate considerable decrease in serum T sub 3 /sub T sub 4 /sub and prolactin levels but no alteration of thyroid-stimulating hormone levels