Learn More
Genscript Corporation CHO-K1/VISTA Stable Cell Line
Supplier:  Genscript Corporation M00533
Encompass
Recombinant CHO-K1 cells stably overexpress V-domain Ig suppressor of T cell activation (VISTA) on the cell surface The surface expression of VISTA is validated by FACS analysis This cell line is designed for cell-based binding for screening antibodies binding with VISTA and evaluating target binding affinity
Catalog No. 50-000-57464
Only null left
Add to Cart
Request a Quote