Learn More
Genscript Corporation CHO-K1/VPAC1/G15 Stable Cell
Supplier:  Genscript Corporation M00342
Encompass
The widespread neuropeptide vasoactive intestinal peptide (VIP) has two receptors VPAC1 and VPAC2 The vasoactive intestinal polypeptide receptor VPAC1 is G sub s /sub -coupled GPCRs expressed in the lung prostate peripheral blood leukocytes liver brain small intestine colon heart spleen placenta kidney thymus and testis
Catalog No. 50-000-58642
Add to Cart
Request a Quote