Learn More
Genscript Corporation CHO-K1/VPAC2/G15 Stable Cell
Catalog No. 5000057078
Encompass
Recombinant CHO-K1 cells stably overexpress human vasoactive intestinal peptide receptor 2 (VPAC2) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57078 Supplier Genscript Corporation Supplier No. M00226
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human vasoactive intestinal peptide receptor 2 (VPAC2) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways