Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CHO Medium
GREENER_CHOICE

Catalog No. 670006
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-000-6 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-000-6 Supplier Gibco™ Supplier No. 670006
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A high-performance, complete, animal origin-free (AOF) formulation offering high-production yields without the need for further supplementation.

This medium is formulated to support cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures. CHO Medium can be used in various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO DG44, BHK-21, CHO-K1, CHO
Classification Chemically-defined, Protein-free, Serum-free
Culture Type Mammalian Cell Culture
Form Powder
Product Type Cell Culture Media
Format Pail
Quantity 10 kg
Source Animal origin-free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.