Learn More
Eurofins Discoverx PathHunter CHO-K1 mGPR91 (SUCNR1) β-Arrestin Cell Pool, 2 vials. (93-0589P2)
Catalog No. 502000899
Encompass
The PathHunter mGPR91 (SUCNR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. succinate) based activation of mGPR91 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGPR91 and EA-tagged β-Arrestin-2. Activation of mGPR91 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0899 Supplier Eurofins Discoverx Supplier No. 930589P2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGPR91 (SUCNR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. succinate) based activation of mGPR91 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGPR91 and EA-tagged β-Arrestin-2. Activation of mGPR91 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.