Learn More
Eurofins Discoverx PathHunter CHO-K1 PPARα Protein Interaction Cell Line, 2 vials. (93-0457C2)
Catalog No. 502000563
Encompass
The PathHunter PPARα Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. leukotriene B4) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. The included cell line overexpresses ProLink-tagged PPARα NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of PPARα, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/nhr.
Catalog No. 50-200-0563 Supplier Eurofins Discoverx Supplier No. 930457C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter PPARα Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. leukotriene B4) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. The included cell line overexpresses ProLink-tagged PPARα NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of PPARα, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/nhr.