Learn More
Cygnus Technologies CHO PROTEIN FREE MEDIA ELISAK
Supplier:  Cygnus Technologies CM015
Encompass
CHO Host Cell Proteins, The CHO HCP assay is a two-site immunoenzymetric assay, This is a new CHO HCP assay developed to be more process specific than the generic assay
Catalog No. NC9885676