Learn More
Cayman Chemical ChoArgtrIfluoroACTAte sal 10mg
SDP
Catalog No. 502935653
Encompass_Preferred
A steroid-based cationic lipid; forms a complex with plasmid DNA and decreases plasmid DNA migration in an agarose-gel retardant assay; facilitates transfection of plasmid DNA into H1299 and HeLa cells in the presence or absence of fetal bovine serum; cytotoxic to H1299 cells (IC50 = 88.5 μg/ml)
Catalog No. 50-293-5653 Supplier Cayman Chemical Supplier No. 2594310 MG
A steroid-based cationic lipid; forms a complex with plasmid DNA and decreases plasmid DNA migration in an agarose-gel retardant assay; facilitates transfection of plasmid DNA into H1299 and HeLa cells in the presence or absence of fetal bovine serum; cytotoxic to H1299 cells (IC50 = 88.5 μg/ml)