Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Chocolate Agar
SDP

Catalog No. 23001749 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-749 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-749 Supplier Hardy Diagnostics™ Supplier No. E14
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the non-selective cultivation of fastidious bacteria

  • Especially useful for the cultivation of Haemophilus and Neisseria in a CO2 enriched atmosphere
  • Contains a highly enriched blend of GC Agar Base, 1% bovine powdered hemoglobin, and 1% MaXV (koenzyme) vitamins and cofactor supplements
  • Optically clear polystyrene for distortion free viewing

Specifications

Appearance Opaque and brown in color
Color Brown
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Dextrose, L-cysteine, HCl, L-glutamine, L-cystine, Adenine, Nicotinic Adenine Dinucleotide, Cocarboxylase, Guanine Hydrochloride, Ferric Nitrate, P-aminobenzoic Acid, Vitamin B12, Thiamine
Detects Fastidious Microorganisms, Haemophilus species, Neisseria species
Form Prepared Medium
Format Agar Plate
Manufacturing Origin United States
Media Type Chocolate Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Fastidious Organisms
Test Type Microbial Cultivation
Fill Volume 19 mL
For Use With (Application) Cultivation of fastidious bacteria, Isolation of fastidious microorganisms
Incubation Conditions 35°C to 37°C
Incubation Time 24 to 48 hr.
Material Polystyrene
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Haemophilus species, Neisseria species
Protein Source Bovine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.