Learn More
Alkali Scientific Chocolate Agar with Enrichment, 20 mL Plate, 10/Pk
SDP
Catalog No. 5000662324
Encompass_Preferred
High quality, enriched Chocolate Agar plates designed for the isolation and cultivation of fastidious organisms such as Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis. Each 90–100 mm plate contains 20 mL of media prepared with lysed red blood cells to release critical growth factors (hemin and NAD), supporting reliable and consistent growth of demanding pathogens. Format: Pre poured 20 mL agar plates Pack : 10 plates per pack Use Case: Ideal for clinical microbiology, diagnostic testing, and research applications Enrichment: Contains X (hemin) and V (NAD) factors naturally released from lysed red blood cells Storage Conditions: Store at 2–8°C; avoid freezing Sterile and ready to use Recommended For: • Culturing respiratory, CSF, and urogenital specimens • Recovery of Haemophilus and Neisseria species • Clinical and reference laboratories
Catalog No. 50-006-62324 Supplier Alkali Scientific Supplier No. CHOCP00
High quality, enriched Chocolate Agar plates designed for the isolation and cultivation of fastidious organisms such as Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis. Each 90–100 mm plate contains 20 mL of media prepared with lysed red blood cells to release critical growth factors (hemin and NAD), supporting reliable and consistent growth of demanding pathogens. Format: Pre poured 20 mL agar plates Pack : 10 plates per pack Use Case: Ideal for clinical microbiology, diagnostic testing, and research applications Enrichment: Contains X (hemin) and V (NAD) factors naturally released from lysed red blood cells Storage Conditions: Store at 2–8°C; avoid freezing Sterile and ready to use Recommended For: • Culturing respiratory, CSF, and urogenital specimens • Recovery of Haemophilus and Neisseria species • Clinical and reference laboratories