Learn More
Thermo Scientific™ Chocolate Agar Medium
Description
Cultivate and isolate fastidious organisms especially Neisseria spp., Haemophilus, and Gardnerella vaginalis using Thermo Scentific™ Chocolate Agar.
Neisseria are capnophilic, gram-negative diplococci or rods that occur in pairs or short chains and inhabit the mucous membranes of humans and animals. Neisseria gonorrhoeae and Neisseria meningitidis are two important pathogens in this group. Gonorrhea is a sexually transmitted disease caused by N. gonorrhoeae and is one of the most frequently reported infectious diseases in the United States and worldwide. This organism is oxidase-positive, and growth is best obtained on Chocolate Agar or Thayer Martin Agar. The treatment and control of gonorrhea is complicated by increasing resistance to a variety of antimicrobial agents, including penicillin, tetracycline, and fluoroquinolones.
Haemophilus are small, pleomorphic, gram-negative coccobacilli. They are nonmotile, non-sporeforming, fastidious, facultative anaerobes. Almost all species require preformed growth factors present in the blood, particularly the X factor (hemin) and/or V factor (NAD) for growth.
Gardnerella vaginalis is found in the genitourinary tract of females and is an indicator organism for bacterial vaginosis (BV). BV is a risk factor for premature labor, chorioamnionitis, and postpartum endometritis. Women with postpartum fever and neonates with sepsis may be infected with Gardnerella vaginalis. In males, Gardnerella vaginalis has been associated with urinary tract infections, prostatitis, and urethritis. Gardnerella vaginalis is a gram-variable rod or coccobacillus. Beta hemolysis is observed on human or rabbit blood-containing media, but not on Sheep Blood Agar.
Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Format | 10mm x 100mm Spacesaver Plate |
| Manufacturing Origin | United States |
| Media Type | Chocolate Agar; Enriched |
| Product Type | Agar Plate |
| Quantity | 150/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.