Learn More
Teknova Chocolate Agar Plates. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388001
Encompass
3.6% GC Agar Base, 1.0% IsoVitalex Enrichment,1.0% Hemoglobin,1.5% Agar
Catalog No. 50-238-8001 Supplier Teknova Supplier No. C49004PK
3.6% GC Agar Base, 1.0% IsoVitalex Enrichment,1.0% Hemoglobin,1.5% Agar