Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Choerospondin | 81202-36-0 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN69635MG

Encompass_Preferred

Choerospondin is a flavanone isolated from the bark of Choerospondias axillaris. It is intended for research use only.

  • Purity of 99.87%
  • Molecular weight of 434.39
  • Molecular formula C21H22O10
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL (230.21 mM)
  • Classified as a flavonoid, flavanone, phenol, and polyphenol
  • Isolated from plants such as Leguminosae - Glycyrrhiza uralensis Fisch. and Anacardiaceae - Choerospondias axillaris

Catalog No. 50-004-19994


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.