Learn More
Science First™ Choice Chamber
Description
- Explore habitat preference and animal behavior using small arthropods as test subjects
- Isopods or “pill bugs” are a good choice
- Chamber is equipped with a tight-fitting lid to contain the critters
- Escape-proof passageway connects the two chambers
- With lid
- Good for AP Biology
Specifications
Specifications
| Product Type | Choice Chamber |
| For Use With (Application) | Study the habits of picky arthropods, explore habitat preference and animal behavior using small arthropods as test subjects |
| Includes | Tight-fitting lid to contain the critters, escape-proof passageway |