Learn More
Thomas Scientific ACTaqT DNA Taq Polymerase, 250Units/Vial
SDP
Catalog No. 501459110
Encompass_Preferred
p> ACTaq™ DNA Polymerase originates from Thermus Aquaticus with molecular weight of 94KDa. For use in DNA fragment extension amplification reactions with DNA having dA overhang on 3’ ends; at extension amplification speed of 1200 bases per minute and optimized between 65-75oC. Optimized for best performance when using dNTP concentration at 100-300mM, 1.5-3.0mM Mg2+ concentration, and pH at 8.1-9.2.
Catalog No. 50-145-9110 Supplier Thomas Scientific Supplier No. C995K08
p> ACTaq™ DNA Polymerase originates from Thermus Aquaticus with molecular weight of 94KDa. For use in DNA fragment extension amplification reactions with DNA having dA overhang on 3’ ends; at extension amplification speed of 1200 bases per minute and optimized between 65-75oC. Optimized for best performance when using dNTP concentration at 100-300mM, 1.5-3.0mM Mg2+ concentration, and pH at 8.1-9.2.