Learn More
Eurofins Discoverx PathHunter CHO-K1 ADORA3 β-Arrestin Cell Line, 2 vials. (93-0586C2)
Supplier:  Eurofins Discoverx 930586C2
Encompass
The PathHunter ADORA3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MECA) based activation of ADORA3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADORA3 and EA-tagged β-Arrestin-2. Activation of ADORA3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0887
Add to Cart
Request a Quote