Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 ADORA3 Gi Cell Line, 2 vials. (95-0101C2)
Catalog No. 502003022
Encompass
The cAMP Hunter ADORA3 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. adenosine, NECA) based activation of ADORA3 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADORA3, a naturally Gi/Go coupled GPCR. Upon activation of ADORA3, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3022 Supplier Eurofins Discoverx Supplier No. 950101C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter ADORA3 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. adenosine, NECA) based activation of ADORA3 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADORA3, a naturally Gi/Go coupled GPCR. Upon activation of ADORA3, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.