Learn More
Eurofins Discoverx CHO-K1 ADRA1A Gq Cell Line, 2 vials. (95-1023C2)
Catalog No. 502004124
Encompass
The ADRA1A Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1A, a naturally Gq coupled GPCR. Upon activation of ADRA1A, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4124 Supplier Eurofins Discoverx Supplier No. 951023C2
Add to Cart
Add to Cart
Request a Quote
The ADRA1A Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1A, a naturally Gq coupled GPCR. Upon activation of ADRA1A, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.