Learn More
Eurofins Discoverx PathHunter CHO-K1 ADRA2A β-Arrestin Cell line, 2 vials. (93-0424C2)
Catalog No. 502000513
Encompass
The PathHunter ADRA2A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2A and EA-tagged β-Arrestin-2. Activation of ADRA2A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0513 Supplier Eurofins Discoverx Supplier No. 930424C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter ADRA2A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2A and EA-tagged β-Arrestin-2. Activation of ADRA2A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.