Learn More
Eurofins Discoverx PathHunter CHO-K1 ADRA2B β-Arrestin Cell Line, 2 vials. (93-0641C2)
Supplier:  Eurofins Discoverx 930641C2
Encompass
The PathHunter ADRA2B cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2B and EA-tagged β-Arrestin-2. Activation of ADRA2B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1013
Add to Cart
Request a Quote