Learn More
Eurofins Discoverx PathHunter CHO-K1 ADRA2C β-Arrestin Cell line, 2 vials. (93-0511C2)
Supplier:  Eurofins Discoverx 930511C2
Encompass
The PathHunter ADRA2C cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2C GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2C and EA-tagged β-Arrestin-2. Activation of ADRA2C stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0681
Only null left
Add to Cart
Request a Quote