Learn More
Eurofins Discoverx PathHunter CHO-K1 BAI3 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0547C2)
Catalog No. 502000773
Encompass
The PathHunter BAI3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of BAI3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged BAI3 and EA-tagged β-Arrestin-2. Activation of BAI3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0773 Supplier Eurofins Discoverx Supplier No. 930547C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter BAI3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of BAI3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged BAI3 and EA-tagged β-Arrestin-2. Activation of BAI3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.