Learn More
Eurofins Discoverx PathHunter CHO-K1 CALCR-RAMP2 (AMY2) β-Arrestin Cell Line, 2 vials. (93-0267C2)
Supplier:  Eurofins Discoverx 930267C2
Encompass
The PathHunter CALCR-RAMP2 (AMY2) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. calcitonin) based activation of CALCR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CALCR and EA-tagged β-Arrestin-2. Activation of CALCR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0125
Add to Cart
Request a Quote