Learn More
Eurofins Discoverx PathHunter CHO-K1 CCKAR β-Arrestin Cell Line, 2 vials. (93-0190C2)
Catalog No. 501999898
Encompass
The PathHunter CCKAR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKAR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCKAR and EA-tagged β-Arrestin-2. Activation of CCKAR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9898 Supplier Eurofins Discoverx Supplier No. 930190C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CCKAR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKAR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCKAR and EA-tagged β-Arrestin-2. Activation of CCKAR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.