Learn More
Eurofins Discoverx PathHunter CHO-K1 CCKBR β-Arrestin Cell Line, 2 vials. (93-0191C2)
Catalog No. 501999902
Encompass
The PathHunter CCKBR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKBR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCKBR and EA-tagged β-Arrestin-2. Activation of CCKBR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9902 Supplier Eurofins Discoverx Supplier No. 930191C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CCKBR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of CCKBR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCKBR and EA-tagged β-Arrestin-2. Activation of CCKBR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.