Learn More
Eurofins Discoverx PathHunter CHO-K1 CCR1 β-Arrestin Cell Line, 2 vials. (93-0254C2)
Catalog No. 502000091
Encompass
The PathHunter CCR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL3) based activation of CCR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR1 and EA-tagged β-Arrestin-2. Activation of CCR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0091 Supplier Eurofins Discoverx Supplier No. 930254C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CCR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL3) based activation of CCR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR1 and EA-tagged β-Arrestin-2. Activation of CCR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.