Learn More
Eurofins Discoverx PathHunter CHO-K1 CCR6 β-Arrestin Cell Line, 2 vials. (93-0194C2)
Supplier: Eurofins Discoverx 930194C2
The PathHunter CCR6 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL20/MIP-3α) based activation of CCR6 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR6 and EA-tagged β-Arrestin-2. Activation of CCR6 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.