Learn More
Eurofins Discoverx PathHunter CHO-K1 cEDG5 β-Arrestin Cell Line, 2 vials. (93-0581C2)
Supplier:  Eurofins Discoverx 930581C2
Encompass
The PathHunter cEDG5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. S1P) based activation of cEDG5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cEDG5 and EA-tagged β-Arrestin-2. Activation of cEDG5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0884
Add to Cart
Request a Quote