Learn More
Eurofins Discoverx PathHunter CHO-K1 CHRM5 β-Arrestin Cell Line, 2 vials. (93-0519C2)
Catalog No. 502000700
Encompass
The PathHunter CHRM5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. oxotremorine) based activation of CHRM5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CHRM5 and EA-tagged β-Arrestin-2. Activation of CHRM5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0700 Supplier Eurofins Discoverx Supplier No. 930519C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CHRM5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. oxotremorine) based activation of CHRM5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CHRM5 and EA-tagged β-Arrestin-2. Activation of CHRM5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.