Learn More
Eurofins Discoverx PathHunter CHO-K1 CNR2 β-Arrestin Cell Line, 2 vials. (93-0706C2)
Supplier:  Eurofins Discoverx 930706C2
Encompass
The PathHunter CNR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CP 55,940) based activation of CNR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CNR2 and EA-tagged β-Arrestin-2. Activation of CNR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1163
Add to Cart
Request a Quote