Learn More
Eurofins Discoverx PathHunter CHO-K1 CRTH2 β-Arrestin Cell Line, 2 vials. (93-0291C2)
Catalog No. 502000182
Encompass
The PathHunter CRTH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of CRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CRTH2 and EA-tagged β-Arrestin-2. Activation of CRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0182 Supplier Eurofins Discoverx Supplier No. 930291C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CRTH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of CRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CRTH2 and EA-tagged β-Arrestin-2. Activation of CRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.