Learn More
Eurofins Discoverx PathHunter CHO-K1 cyno GPR120 β-Arrestin Cell Line, 2 vials. (93-1029C2)
Catalog No. 502001880
Encompass
The PathHunter cyno GPR120 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of cyno GPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno GPR120 and EA-tagged β-Arrestin-2. Activation of cyno GPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1880 Supplier Eurofins Discoverx Supplier No. 931029C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter cyno GPR120 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of cyno GPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno GPR120 and EA-tagged β-Arrestin-2. Activation of cyno GPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.