Learn More
Eurofins Discoverx CHO-K1 CYSLTR2 Gq Cell Line, 2 vials. (95-1013C2)
Supplier:  Eurofins Discoverx 951013C2
Encompass
The CYSLTR2 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene) based activation of CYSLTR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CYSLTR2, a naturally Gq coupled GPCR. Upon activation of CYSLTR2, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4073
Only null left
Add to Cart
Request a Quote