Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 DRD1 Gs Cell Line, 2 vials. (95-0083C2)
Supplier:  Eurofins Discoverx 950083C2
Encompass
The cAMP Hunter DRD1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type DRD1, a naturally Gs coupled GPCR. Upon activation of DRD1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2785
Only null left
Add to Cart
Request a Quote