Learn More
Eurofins Discoverx PathHunter CHO-K1 DRD2L (Long Isoform) β-Arrestin Cell Line, 2 vials. (93-0579C2)
Catalog No. 502000876
Encompass
The PathHunter DRD2L cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD2L GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD2L and EA-tagged β-Arrestin-2. Activation of DRD2L stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0876 Supplier Eurofins Discoverx Supplier No. 930579C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter DRD2L cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD2L GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD2L and EA-tagged β-Arrestin-2. Activation of DRD2L stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.