Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 DRD2L (Long Isoform) Gi Cell Line, 2 vials. (95-0125C2)
Supplier:  Eurofins Discoverx 950125C2
Encompass
The cAMP Hunter DRD2L Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD2L GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type DRD2L, a naturally Gi/Go coupled GPCR. Upon activation of DRD2L, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3355
Add to Cart
Request a Quote