Learn More
Eurofins Discoverx PathHunter CHO-K1 DRD4 β-Arrestin Cell Line, 2 vials. (93-0690C2)
Catalog No. 502001115
Encompass
The PathHunter DRD4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD4 and EA-tagged β-Arrestin-2. Activation of DRD4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1115 Supplier Eurofins Discoverx Supplier No. 930690C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter DRD4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD4 and EA-tagged β-Arrestin-2. Activation of DRD4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.