Learn More
Eurofins Discoverx PathHunter CHO-K1 EDG4 β-Arrestin Cell Line, 2 vials. (93-0255C2)
Catalog No. 502000092
Encompass
The PathHunter EDG4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of EDG4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG4 and EA-tagged β-Arrestin-2. Activation of EDG4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0092 Supplier Eurofins Discoverx Supplier No. 930255C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter EDG4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of EDG4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG4 and EA-tagged β-Arrestin-2. Activation of EDG4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.