Learn More
Eurofins Discoverx DNA 12K Reagent Kit
Catalog No. 502000000
Encompass
DNA Reagent for the DNA 12K LabChip kit. Contains enough reagent for 2000 wells. The kit for use on the LabChip GX and LabChip GX Touch systems provides a fast and easy way to analyze a DNA samples ranging from 100 to 12,000 base pairs, which is an extended size range relative to the companion assays in the PerkinElmer portfolio. Performing DNA sample analysis with the DNA 12K assay saves time and money by automating steps for analyzing size and purity, and replaces agarose slab gels, and imaging. The LabChip GX and GX Touch Electrophoresis Systems uses a single sipper microfluidic chip to aspirate dsDNA samples directly from 96- or 384-well plates. Additional Specifications Sizing Range - 100 bp – 12000 bp Sizing Accuracy - ± 10% Sizing Resolution - ± 10% from 150 – 1000 bp ± 15% from 1000 – 2000 bp ± 20% from 2000 – 8000 bp ± 25% from 100 – 150 bp, 8000 – 12000 bp Linear Concentration Range – 0.25 ng/µL – 50 ng / µL Per fragment Sensitivity – 0.25 ng / µL Chip Sample Lifetime –2000
Catalog No. 50-200-0000 Supplier Eurofins Discoverx Supplier No. 930227C2
Add to Cart
Add to Cart
Request a Quote
DNA Reagent for the DNA 12K LabChip kit. Contains enough reagent for 2000 wells. The kit for use on the LabChip GX and LabChip GX Touch systems provides a fast and easy way to analyze a DNA samples ranging from 100 to 12,000 base pairs, which is an extended size range relative to the companion assays in the PerkinElmer portfolio. Performing DNA sample analysis with the DNA 12K assay saves time and money by automating steps for analyzing size and purity, and replaces agarose slab gels, and imaging. The LabChip GX and GX Touch Electrophoresis Systems uses a single sipper microfluidic chip to aspirate dsDNA samples directly from 96- or 384-well plates. Additional Specifications Sizing Range - 100 bp – 12000 bp Sizing Accuracy - ± 10% Sizing Resolution - ± 10% from 150 – 1000 bp ± 15% from 1000 – 2000 bp ± 20% from 2000 – 8000 bp ± 25% from 100 – 150 bp, 8000 – 12000 bp Linear Concentration Range – 0.25 ng/µL – 50 ng / µL Per fragment Sensitivity – 0.25 ng / µL Chip Sample Lifetime –2000