Learn More
Eurofins Discoverx PathHunter CHO-K1 EDG7 β-Arrestin Cell Line, 2 vials. (93-0636C2)
Catalog No. 502000997
Encompass
The PathHunter EDG7 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of EDG7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG7 and EA-tagged β-Arrestin-2. Activation of EDG7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0997 Supplier Eurofins Discoverx Supplier No. 930636C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter EDG7 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of EDG7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG7 and EA-tagged β-Arrestin-2. Activation of EDG7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.